Assay Details
| Gene Symbol: Arr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133205.3 | NP_573468.1 | 2 - 3 | 114 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133205.3 transcripts.
Set Membership: |
|

Arr3
Genomic Map