Assay Details
| Gene Symbol: Igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008784.3 | NP_032810.4 | 3 - 4 | 751 | 106 | |
| GenBank mRNA | AA213189.1 | - | 2 - 3 | 278 | 106 | |
| BC089567.1 | - | 3 - 4 | 683 | 30297519 | 106 | |
| CX567459.1 | - | 2 - 3 | 231 | 106 | ||
| L31652.1 | - | 3 - 4 | 676 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igbp1
Genomic Map