Assay Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290801.1 | NP_001277730.1 | 13 - 14 | 1714 | 83 | |
| NM_173870.3 | NP_776295.1 | 14 - 15 | 1793 | 83 | ||
| GenBank mRNA | AB053217.1 | - | 13 - 14 | 1469 | 83 | |
| AK052278.1 | - | 11 - 12 | 1290 | 83 | ||
| AK155724.1 | - | 14 - 15 | 1795 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgat4a
Genomic Map