Assay Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290801.1 | NP_001277730.1 | 6 - 7 | 940 | 63 | |
| NM_173870.3 | NP_776295.1 | 7 - 8 | 1019 | 63 | ||
| GenBank mRNA | AB053217.1 | - | 6 - 7 | 695 | 63 | |
| AK052278.1 | - | 4 - 5 | 516 | 63 | ||
| AK155724.1 | - | 7 - 8 | 1020 | 63 | ||
| AK163327.1 | - | 6 - 7 | 772 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgat4a
Genomic Map