Assay Details
| Gene Symbol: Eif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198303.2 | NP_938045.2 | 10 - 11 | 2107 | 105 | |
| GenBank mRNA | BC103806.1 | - | 9 - 10 | 2860 | 105 | |
| CN670500.1 | - | 1 - 2 | 140 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif5b
Genomic Map