Assay Details
| Gene Symbol: Eif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198303.2 | NP_938045.2 | 18 - 19 | 3145 | 119 | |
| GenBank mRNA | AK037164.1 | - | 2 - 3 | 252 | 119 | |
| AK173016.1 | - | 2 - 3 | 218 | 119 | ||
| BC055393.1 | - | 2 - 3 | 223 | 119 | ||
| BC103806.1 | - | 17 - 18 | 3898 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif5b
Genomic Map