Assay Details
| Gene Symbol: Eif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198303.2 | NP_938045.2 | 20 - 21 | 3362 | 76 | |
| GenBank mRNA | AK037164.1 | - | 4 - 5 | 469 | 76 | |
| AK173016.1 | - | 4 - 5 | 435 | 76 | ||
| BC055393.1 | - | 4 - 5 | 440 | 76 | ||
| BC103806.1 | - | 19 - 20 | 4115 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif5b
Genomic Map