Assay Details
| Gene Symbol: Msh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010830.2 | NP_034960.1 | 2 - 3 | 554 | 67 | |
| GenBank mRNA | AK088259.1 | - | 2 - 3 | 543 | 67 | |
| BC051160.1 | - | 2 - 3 | 499 | 4953238 | 67 | |
| BC051634.1 | - | 2 - 3 | 509 | 6518577 | 67 | |
| U42190.1 | - | 2 - 3 | 530 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh6
Genomic Map