Assay Details
| Gene Symbol: Msh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010830.2 | NP_034960.1 | 3 - 4 | 725 | 114 | |
| GenBank mRNA | AK088259.1 | - | 3 - 4 | 714 | 114 | |
| BC035274.1 | - | 1 - 2 | 123 | 114 | ||
| BC051160.1 | - | 3 - 4 | 670 | 4953238 | 114 | |
| BC051634.1 | - | 3 - 4 | 680 | 6518577 | 114 | |
| U42190.1 | - | 3 - 4 | 701 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh6
Genomic Map