Assay Details
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153551.1 | NP_705779.1 | 13 - 14 | 1034 | 75 | |
GenBank mRNA | AK040537.1 | - | 13 - 14 | 1052 | 75 | |
BC035208.1 | - | 13 - 14 | 1034 | 3671384 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
