Assay Details
| Gene Symbol: Terf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286628.1 | NP_001273557.1 | 4 - 5 | 685 | 137 | |
| NM_009352.3 | NP_033378.1 | 5 - 6 | 772 | 137 | ||
| GenBank mRNA | AB109563.1 | - | 5 - 6 | 752 | 137 | |
| AK021235.1 | - | 5 - 6 | 762 | 137 | ||
| AK036615.1 | - | 5 - 6 | 775 | 137 | ||
| AK132022.1 | - | 4 - 5 | 668 | 137 | ||
| BC034866.1 | - | 5 - 6 | 760 | 3470277 | 137 | |
| U65586.1 | - | 5 - 6 | 752 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Terf1
Genomic Map