Assay Details
| Gene Symbol: Fbxo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081034.1 | NP_001074503.1 | 17 - 18 | 2129 | 84 | |
| GenBank mRNA | AK132635.1 | - | 1 - 2 | 105 | 84 | |
| BC027802.1 | - | 6 - 7 | 706 | 84 | ||
| BC049946.1 | - | 1 - 2 | 57 | 84 | ||
| BC055343.1 | - | 8 - 9 | 871 | 84 | ||
| BC110686.1 | - | 17 - 18 | 1868 | 84 | ||
| BC117884.1 | - | 17 - 18 | 1875 | 40096054 | 84 | |
| BC117885.1 | - | 17 - 18 | 1879 | 40096055 | 84 | |
| BC128479.1 | - | 17 - 18 | 1879 | 40096053 | 84 | |
More Information
Set Membership: |
|

Fbxo11
Genomic Map