Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700010502.mm.1 |
NM_001128181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010502.mm.1 |
NM_001128181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010502.mm.1 |
NM_177639.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010502.mm.1 |
NM_177639.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010502.mm.1 |
TC1700001113.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_001128180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_001128180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_001128181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_001128181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_027712.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_027712.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_177639.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
NM_177639.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010500.mm.1 |
TC1700001113.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_001128180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_001128180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_001128181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_001128181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_027712.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_027712.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_177639.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
NM_177639.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010504.mm.1 |
TC1700001113.mm.1 |
MTA TC |