Assay Details
Gene Symbol: Sgk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037759.1 | NP_001032848.1 | 12 - 13 | 1116 | 78 | |
NM_133220.2 | NP_573483.1 | 13 - 14 | 1235 | 78 | ||
NM_177547.3 | NP_808215.2 | 12 - 13 | 1128 | 78 | ||
GenBank mRNA | AF312007.1 | - | 13 - 14 | 1177 | 78 | |
AK028722.1 | - | 13 - 14 | 1180 | 78 | ||
AK030314.1 | - | 12 - 13 | 1117 | 78 | ||
AK031133.1 | - | 13 - 14 | 1216 | 78 | ||
AK031328.1 | - | 12 - 13 | 1057 | 78 | ||
AK083319.1 | - | 3 - 4 | 183 | 78 | ||
AK143836.1 | - | 13 - 14 | 1223 | 78 | ||
AK146561.1 | - | 12 - 13 | 1071 | 78 | ||
AK151181.1 | - | 13 - 14 | 1180 | 78 | ||
AK151793.1 | - | 12 - 13 | 1108 | 78 | ||
AK151867.1 | - | 13 - 14 | 1212 | 78 | ||
AK151996.1 | - | 13 - 14 | 1379 | 78 | ||
AK166414.1 | - | 12 - 13 | 1058 | 78 | ||
AK166531.1 | - | 12 - 13 | 1077 | 78 | ||
AK171120.1 | - | 12 - 13 | 1104 | 78 | ||
AK171186.1 | - | 12 - 13 | 1104 | 78 | ||
AK171678.1 | - | 12 - 13 | 1088 | 78 | ||
AK171959.1 | - | 12 - 13 | 1165 | 78 | ||
AK172323.1 | - | 12 - 13 | 1078 | 78 | ||
AK196812.1 | - | 2 - 3 | 88 | 78 | ||
BC018363.1 | - | 10 - 11 | 766 | 2631682 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
