Assay Details
Gene Symbol: Ankrd13d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026720.2 | NP_080996.2 | 3 - 4 | 517 | 78 | |
GenBank mRNA | AK089186.1 | - | 3 - 4 | 510 | 78 | |
AK155698.1 | - | 3 - 4 | 518 | 78 | ||
BC058982.1 | - | 3 - 4 | 428 | 5685059 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
