Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900007971.mm.1 |
NM_026720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007971.mm.1 |
NM_026720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007971.mm.1 |
TC1900000915.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007973.mm.1 |
NM_026720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007973.mm.1 |
NM_026720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007973.mm.1 |
TC1900000915.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007970.mm.1 |
NM_026720.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007970.mm.1 |
NM_026720.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007970.mm.1 |
TC1900000915.mm.1 |
MTA TC |