Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100017810.mm.1 |
NM_026913.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017810.mm.1 |
NM_026913.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017810.mm.1 |
TC0100002210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017813.mm.1 |
NM_026913.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017813.mm.1 |
NM_026913.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017813.mm.1 |
TC0100002210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017811.mm.1 |
NM_026913.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017811.mm.1 |
NM_026913.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017811.mm.1 |
TC0100002210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100017812.mm.1 |
NM_026913.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017812.mm.1 |
NM_026913.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100017812.mm.1 |
TC0100002210.mm.1 |
MTA TC |