Assay Details
| Gene Symbol: Amer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175179.4 | NP_780388.2 | 2 | 68 | ||
| GenBank mRNA | AK049388.1 | - | 1 | 68 | ||
| BC060714.1 | - | 1 | 68 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Amer1
Genomic Map