Assay Details
Gene Symbol: Cebpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024806.2 | NP_001019977.1 | 11 - 12 | 2649 | 94 | |
GenBank mRNA | AK136173.1 | - | 11 - 12 | 2630 | 94 | |
AK145374.1 | - | 11 - 12 | 2631 | 94 | ||
AK155990.1 | - | 11 - 12 | 2632 | 94 | ||
AK158771.1 | - | 11 - 12 | 2626 | 94 | ||
AK159081.1 | - | 11 - 12 | 2646 | 94 | ||
BB704756.1 | - | 3 - 4 | 239 | 94 | ||
U19891.1 | - | 11 - 12 | 2620 | 94 | ||
U19892.1 | - | 11 - 12 | 2334 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
