Assay Details
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290729.1 | NP_001277658.1 | 14 - 15 | 2376 | 110 | |
| GenBank mRNA | AB299229.1 | - | 14 - 15 | 2303 | 110 | |
| BC047418.1 | - | 4 - 5 | 489 | 110 | ||
| BC051073.1 | - | 1 - 2 | 102 | 110 | ||
More Information
Set Membership: |
|

Taf1
Genomic Map