Assay Details
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290729.1 | NP_001277658.1 | 18 - 19 | 2919 | 118 | |
| GenBank mRNA | AB299229.1 | - | 18 - 19 | 2846 | 118 | |
| AF022178.1 | - | 3 - 4 | 234 | 118 | ||
| AK050691.1 | - | 3 - 4 | 290 | 118 | ||
| BC047418.1 | - | 8 - 9 | 1032 | 118 | ||
| BC051073.1 | - | 5 - 6 | 645 | 118 | ||
More Information
Set Membership: |
|

Taf1
Genomic Map