Assay Details
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290729.1 | NP_001277658.1 | 2 - 3 | 379 | 70 | |
| GenBank mRNA | AB299229.1 | - | 2 - 3 | 306 | 70 | |
| AK045586.2 | - | 2 - 3 | 378 | 70 | ||
| AK143571.1 | - | 2 - 3 | 356 | 70 | ||
More Information
Set Membership: |
|

Taf1
Genomic Map