Assay Details
Gene Symbol: 4933400B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045432.1 | 4 - 5 | 579 | 106 | ||
GenBank mRNA | AK016587.1 | - | 4 - 5 | 580 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | follicle stimulating hormone receptor | Fshr | -125.5kb | Anti-sense | Chr. 17 - 88984952 - 89200675 on NCBI Build GRCm38 | |
Downstream | predicted gene 4719 | Gm4719 | 51.9kb | Sense | Chr. 17 - 89379241 - 89384993 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
