Assay Details
| Gene Symbol: Arid5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172205.1 | NP_001165676.1 | 3 - 4 | 350 | 60 | |
| NM_001172206.1 | NP_001165677.1 | 3 - 4 | 347 | 60 | ||
| NM_001290726.1 | NP_001277655.1 | 4 - 5 | 445 | 60 | ||
| GenBank mRNA | AK149839.1 | - | 3 - 4 | 330 | 60 | |
| AK150170.1 | - | 4 - 5 | 425 | 60 | ||
| AK156376.1 | - | 3 - 4 | 330 | 60 | ||
| AK157607.1 | - | 3 - 4 | 330 | 60 | ||
More Information
Set Membership: |
|

Arid5a
Genomic Map