Assay Details
| Gene Symbol: Sema4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148298.1 | - | 10 - 11 | 1350 | 76 | |
| AK173247.1 | - | 11 - 12 | 1590 | 76 | ||
| S79463.1 | - | 10 - 11 | 1204 | 76 | ||
More Information
Set Membership: |
|

Sema4c
Genomic Map