Assay Details
| Gene Symbol: Bfsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002896.2 | NP_001002896.1 | 5 - 6 | 1125 | 61 | |
| GenBank mRNA | AK134810.1 | - | 5 - 6 | 1057 | 61 | |
| AK178846.1 | - | 1 - 2 | 109 | 61 | ||
| AK195636.1 | - | 1 - 2 | 109 | 61 | ||
| BC062815.1 | - | 3 - 4 | 327 | 61 | ||
| BC068172.1 | - | 5 - 6 | 1125 | 30550909 | 61 | |
| S55549.1 | - | 3 - 4 | 434 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bfsp2
Genomic Map