Assay Details
| Gene Symbol: Mst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008243.3 | NP_032269.3 | 5 - 6 | 608 | 62 | |
| GenBank mRNA | AK040934.1 | - | 5 - 6 | 592 | 62 | |
| AK134222.1 | - | 5 - 6 | 591 | 62 | ||
| AK144824.1 | - | 5 - 6 | 571 | 62 | ||
| BC010551.1 | - | 5 - 6 | 593 | 4195560 | 62 | |
| BC051393.1 | - | 5 - 6 | 577 | 5055055 | 62 | |
| M74181.1 | - | 5 - 6 | 565 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mst1
Genomic Map