Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900010751.mm.1 |
NM_018879.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010751.mm.1 |
NM_018879.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010751.mm.1 |
TC0900001371.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010752.mm.1 |
NM_018879.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010752.mm.1 |
NM_018879.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010752.mm.1 |
TC0900001371.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010753.mm.1 |
NM_018879.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010753.mm.1 |
NM_018879.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010753.mm.1 |
TC0900001371.mm.1 |
MTA TC |