Assay Details
Gene Symbol: 1700128A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045938.2 | 1 - 2 | 119 | 76 | ||
GenBank mRNA | AK007305.1 | - | 1 - 2 | 116 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
