Assay Details
| Gene Symbol: Uba5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025692.3 | NP_079968.2 | 5 - 6 | 666 | 140 | |
| GenBank mRNA | AK077825.1 | - | 5 - 6 | 647 | 140 | |
| AK151823.1 | - | 5 - 6 | 623 | 140 | ||
| AK152029.1 | - | 5 - 6 | 661 | 140 | ||
| AK168907.1 | - | 5 - 6 | 660 | 140 | ||
| BC019764.1 | - | 5 - 6 | 543 | 3989119 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uba5
Genomic Map