Assay Details
| Gene Symbol: Srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009275.4 | NP_033301.1 | 2 - 3 | 245 | 72 | |
| GenBank mRNA | AK004854.1 | - | 2 - 3 | 278 | 72 | |
| AK007859.1 | - | 2 - 3 | 251 | 72 | ||
| AK008383.1 | - | 2 - 3 | 288 | 72 | ||
| AK015469.1 | - | 2 - 3 | 310 | 72 | ||
| AK030437.1 | - | 2 - 3 | 246 | 72 | ||
| AK075583.1 | - | 2 - 3 | 248 | 72 | ||
| AK160600.1 | - | 2 - 3 | 311 | 72 | ||
| AK167039.1 | - | 2 - 3 | 251 | 72 | ||
| BC003798.1 | - | 2 - 3 | 266 | 3490766 | 72 | |
| U17343.1 | - | 2 - 3 | 259 | 72 | ||
More Information
Set Membership: |
|

Srprb
Genomic Map