Assay Details
Gene Symbol: Trf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133977.2 | NP_598738.1 | 3 - 4 | 398 | 91 | |
GenBank mRNA | AF440692.1 | - | 2 - 3 | 328 | 91 | |
AK085754.1 | - | 3 - 4 | 383 | 91 | ||
AK142599.1 | - | 388 | 91 | |||
AK146549.1 | - | 3 - 4 | 383 | 91 | ||
AK149559.1 | - | 3 - 4 | 383 | 91 | ||
AK149595.1 | - | 3 - 4 | 383 | 91 | ||
AK150782.1 | - | 3 - 4 | 431 | 91 | ||
AK168368.1 | - | 3 - 4 | 383 | 91 | ||
AK168405.1 | - | 3 - 4 | 383 | 91 | ||
AK168419.1 | - | 3 - 4 | 383 | 91 | ||
AK168938.1 | - | 3 - 4 | 383 | 91 | ||
AK169187.1 | - | 3 - 4 | 383 | 91 | ||
BC008559.1 | - | 1 - 2 | 120 | 91 | ||
BC012313.1 | - | 3 - 4 | 398 | 3981648 | 91 | |
BC020295.1 | - | 3 - 4 | 367 | 91 | ||
BC022986.1 | - | 3 - 4 | 376 | 5123741 | 91 | |
BC058216.1 | - | 3 - 4 | 364 | 91 | ||
BC058218.1 | - | 3 - 4 | 377 | 91 | ||
BC092046.1 | - | 3 - 4 | 359 | 3594705 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
