Assay Details
| Gene Symbol: Mapkapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178907.3 | NP_849238.1 | 2 - 3 | 625 | 113 | |
| GenBank mRNA | AK051962.1 | - | 2 - 3 | 496 | 113 | |
| AK087496.1 | - | 1 - 2 | 286 | 113 | ||
| AK144637.1 | - | 2 - 3 | 563 | 113 | ||
| AK151881.1 | - | 2 - 3 | 548 | 113 | ||
| AK170456.1 | - | 2 - 3 | 622 | 113 | ||
| AK172344.1 | - | 2 - 3 | 536 | 113 | ||
| AK172578.1 | - | 2 - 3 | 528 | 113 | ||
| BC031467.1 | - | 2 - 3 | 625 | 4189474 | 113 | |
| DQ899946.1 | - | 1 - 2 | 220 | 113 | ||
More Information
Set Membership: |
|

Mapkapk3
Genomic Map