Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900025299.mm.1 |
NM_133984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025299.mm.1 |
NM_133984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025299.mm.1 |
TC0900003028.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025303.mm.1 |
NM_133984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025303.mm.1 |
NM_133984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025303.mm.1 |
TC0900003028.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025301.mm.1 |
NM_133984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025301.mm.1 |
NM_133984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025301.mm.1 |
TC0900003028.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025302.mm.1 |
NM_133984.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025302.mm.1 |
NM_133984.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025302.mm.1 |
TC0900003028.mm.1 |
MTA TC |