Assay Details
| Gene Symbol: Oprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204371.1 | NP_001191300.1 | 2 - 3 | 478 | 88 | |
| NM_011011.2 | NP_035141.1 | 2 - 3 | 448 | 88 | ||
| GenBank mRNA | BC116795.1 | - | 2 - 3 | 384 | 40126114 | 88 |
| BC119026.1 | - | 2 - 3 | 384 | 8733775 | 88 | |
| BC144731.1 | - | 2 - 3 | 414 | 88 | ||
| L11065.1 | - | 2 - 3 | 448 | 88 | ||
| S81111.1 | - | 2 - 3 | 359 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oprk1
Genomic Map