Assay Details
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026329.2 | NP_080605.1 | 2 - 3 | 210 | 82 | |
GenBank mRNA | AK007892.1 | - | 2 - 3 | 231 | 82 | |
AK010687.1 | - | 2 - 3 | 179 | 82 | ||
AK011627.1 | - | 2 - 3 | 211 | 82 | ||
AK013134.1 | - | 2 - 3 | 181 | 82 | ||
AK017473.1 | - | 2 - 3 | 189 | 82 | ||
BC005580.1 | - | 2 - 3 | 229 | 3968561 | 82 | |
BC055278.1 | - | 2 - 3 | 160 | 5699685 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
