Assay Details
Gene Symbol: Crem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110851.1 | NP_001104321.1 | 2 - 3 | 323 | 73 | |
NM_001110856.2 | NP_001104326.1 | 2 - 3 | 211 | 73 | ||
NM_001110859.2 | NP_001104329.1 | 2 - 3 | 211 | 73 | ||
NM_001271503.1 | NP_001258432.1 | 2 - 3 | 211 | 73 | ||
NM_001271504.1 | NP_001258433.1 | 2 - 3 | 211 | 73 | ||
NM_001271506.1 | NP_001258435.1 | 2 - 3 | 211 | 73 | ||
GenBank mRNA | AK161366.1 | - | 2 - 3 | 257 | 73 | |
AY738715.1 | - | 1 - 2 | 127 | 73 | ||
AY738717.1 | - | 1 - 2 | 127 | 73 | ||
AY738718.1 | - | 1 - 2 | 127 | 73 | ||
AY738721.1 | - | 1 - 2 | 127 | 73 | ||
S64590.1 | - | 1 - 2 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
