Assay Details
| Gene Symbol: Hnrnpul2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081196.1 | NP_001074665.1 | 10 - 11 | 2650 | 87 | |
| GenBank mRNA | DQ000354.1 | - | 10 - 11 | 1769 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpul2
Genomic Map