Assay Details
| Gene Symbol: Hnrnpul2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081196.1 | NP_001074665.1 | 11 - 12 | 2861 | 82 | |
| GenBank mRNA | DQ000354.1 | - | 11 - 12 | 1980 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P18167 | NM_001081196.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P18167 | AK004010.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464779 | AK078249.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386283 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122484 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36987 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P18167 | NM_001081196.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P18167 | AK004010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P308828 | AK032708.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14502905 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14502904 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14502900 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461236 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461222 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434551_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415765_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461220 | - | - |
| Mouse Genome 430 2.0 Array | 1415765_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415765_at | - | - |
| Mouse Genome 430 2.0 Array | 1434551_at | - | - |
| Mouse Genome 430 2.0 Array | 1434551_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68693_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68693_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900002114.mm.1 | NM_001081196.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002114.mm.1 | NM_001081196.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002114.mm.1 | TC1900000159.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434551_at | - | - |
| Mouse Expression Set 430 | 1415765_PM_at | - | - |
| Mouse Expression Set 430 | 1434551_PM_at | - | - |
| Mouse Expression Set 430 | 1415765_at | - | - |
| Mouse Expression Set 430 | 1415765_PM_at | - | - |
| Mouse Expression Set 430 | 1415765_at | - | - |
| Murine Genome U74v2 Set | 104609_PM_at | - | - |
| Murine Genome U74v2 Set | 104609_at | - | - |
| Murine Genome U74v2 Set | 104296_at | - | - |
| Murine Genome U74v2 Set | 104296_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434551_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415765_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434551_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415765_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434551_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415765_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415765_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434551_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415765_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415765_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 504730 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106040575 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247431 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220918 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2666851 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106040575 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106040575 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220918 | - | - |

Hnrnpul2
Genomic Map