Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 16 - 17 | 3385 | 104 | |
| NM_016813.2 | NP_058093.2 | 17 - 18 | 1634 | 104 | ||
| GenBank mRNA | AF093140.1 | - | 17 - 18 | 1610 | 104 | |
| AK052937.1 | - | 7 - 8 | 801 | 104 | ||
| AK077644.1 | - | 17 - 18 | 1591 | 104 | ||
| AK079583.1 | - | 17 - 18 | 1623 | 104 | ||
| AK086527.1 | - | 16 - 17 | 3396 | 104 | ||
| AK088634.1 | - | 16 - 17 | 3356 | 104 | ||
| AK133883.1 | - | 17 - 18 | 1623 | 104 | ||
| AK143640.1 | - | 3386 | 104 | |||
| AK167518.1 | - | 17 - 18 | 1622 | 104 | ||
| BC005594.1 | - | 17 - 18 | 1634 | 3710546 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map