Assay Details
| Gene Symbol: Bscl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136064.3 | NP_001129536.1 | 8 - 9 | 1523 | 57 | |
| NM_001290823.1 | NP_001277752.1 | 7 - 8 | 982 | 57 | ||
| NM_008144.5 | NP_032170.3 | 7 - 8 | 1373 | 57 | ||
| GenBank mRNA | AB030196.1 | - | 7 - 8 | 992 | 57 | |
| AF069954.2 | - | 7 - 8 | 1198 | 57 | ||
| AK004634.1 | - | 5 - 6 | 3072 | 57 | ||
| AK151606.1 | - | 7 - 8 | 951 | 57 | ||
| AK155753.1 | - | 6 - 7 | 1490 | 57 | ||
| AK158901.1 | - | 7 - 8 | 1374 | 57 | ||
| AK170183.1 | - | 7 - 8 | 949 | 57 | ||
| BC043023.1 | - | 7 - 8 | 1137 | 5689013 | 57 | |
| BC060048.1 | - | 6 - 7 | 2203 | 57 | ||
| BC061689.1 | - | 8 - 9 | 1261 | 6306706 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bscl2
Genomic Map