Assay Details
| Gene Symbol: Gng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010316.3 | NP_034446.1 | 1 - 2 | 257 | 99 | |
| GenBank mRNA | AK004459.1 | - | 1 - 2 | 222 | 99 | |
| AK032646.1 | - | 1 - 2 | 205 | 99 | ||
| BC029680.1 | - | 1 - 2 | 141 | 5365311 | 99 | |
| BY290040.1 | - | 1 - 2 | 342 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004459.1, AK032646.1, BC029680.1, NM_010316.3 transcripts.
Set Membership: |
|

Gng3
Genomic Map