Assay Details
| Gene Symbol: Adam17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277266.1 | NP_001264195.1 | 18 - 19 | 2344 | 101 | |
| NM_001291871.1 | NP_001278800.1 | 17 - 18 | 2306 | 101 | ||
| NM_009615.6 | NP_033745.4 | 17 - 18 | 2287 | 101 | ||
| NR_102380.1 | 14 - 15 | 1930 | 101 | |||
| NR_120376.1 | 17 - 18 | 2310 | 101 | |||
| GenBank mRNA | AB021709.1 | - | 17 - 18 | 2248 | 101 | |
| AJ007365.1 | - | 17 - 18 | 2087 | 101 | ||
| AK137693.1 | - | 17 - 18 | 2194 | 101 | ||
| AK139471.1 | - | 17 - 18 | 2193 | 101 | ||
| AK142441.1 | - | 17 - 18 | 2246 | 101 | ||
| AK144161.1 | - | 18 - 19 | 2274 | 101 | ||
| AK149288.1 | - | 17 - 18 | 2190 | 101 | ||
| BC026052.1 | - | 14 - 15 | 1826 | 101 | ||
| BC048792.1 | - | 17 - 18 | 2194 | 101 | ||
| BC094655.1 | - | 17 - 18 | 2201 | 5705503 | 101 | |
| BC114999.1 | - | 17 - 18 | 2147 | 6506501 | 101 | |
| BC138416.1 | - | 17 - 18 | 2166 | 8861436 | 101 | |
| BC138420.1 | - | 17 - 18 | 2166 | 8861440 | 101 | |
| BC145270.1 | - | 17 - 18 | 2166 | 9053824 | 101 | |
| U69613.1 | - | 17 - 18 | 2155 | 101 | ||
| U69614.1 | - | 17 - 18 | 2262 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam17
Genomic Map