Assay Details
| Gene Symbol: Adam17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277266.1 | NP_001264195.1 | 1 - 2 | 295 | 80 | |
| NM_001291871.1 | NP_001278800.1 | 1 - 2 | 295 | 80 | ||
| NM_009615.6 | NP_033745.4 | 1 - 2 | 295 | 80 | ||
| NR_120376.1 | 1 - 2 | 295 | 80 | |||
| GenBank mRNA | AB021709.1 | - | 1 - 2 | 256 | 80 | |
| AJ007365.1 | - | 1 - 2 | 95 | 80 | ||
| AK137693.1 | - | 1 - 2 | 202 | 80 | ||
| AK139471.1 | - | 1 - 2 | 201 | 80 | ||
| AK142441.1 | - | 1 - 2 | 235 | 80 | ||
| AK144161.1 | - | 1 - 2 | 225 | 80 | ||
| AK149288.1 | - | 1 - 2 | 198 | 80 | ||
| AK149614.1 | - | 1 - 2 | 202 | 80 | ||
| AK165739.1 | - | 1 - 2 | 202 | 80 | ||
| BC048792.1 | - | 1 - 2 | 203 | 80 | ||
| BC094655.1 | - | 1 - 2 | 209 | 5705503 | 80 | |
| BC114999.1 | - | 1 - 2 | 155 | 6506501 | 80 | |
| BC138416.1 | - | 1 - 2 | 174 | 8861436 | 80 | |
| BC138420.1 | - | 1 - 2 | 174 | 8861440 | 80 | |
| BC145270.1 | - | 1 - 2 | 174 | 9053824 | 80 | |
| CA325381.1 | - | 1 - 2 | 263 | 80 | ||
| U69613.1 | - | 1 - 2 | 163 | 80 | ||
| U69614.1 | - | 1 - 2 | 247 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291871.1 transcripts.
Set Membership: |
|

Adam17
Genomic Map