Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900007718.mm.1 |
NM_026316.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007718.mm.1 |
NM_026316.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007718.mm.1 |
TC1900000890.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007714.mm.1 |
NM_026316.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007714.mm.1 |
NM_026316.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007714.mm.1 |
TC1900000890.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007717.mm.1 |
NM_026316.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007717.mm.1 |
NM_026316.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007717.mm.1 |
TC1900000890.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007716.mm.1 |
NM_026316.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007716.mm.1 |
NM_026316.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007716.mm.1 |
TC1900000890.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007713.mm.1 |
NM_026316.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007713.mm.1 |
NM_026316.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007713.mm.1 |
TC1900000890.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007715.mm.1 |
NM_026316.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007715.mm.1 |
NM_026316.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007715.mm.1 |
TC1900000890.mm.1 |
MTA TC |