Assay Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039657.2 | NP_001034746.1 | 5 - 6 | 919 | 55 | |
| NM_001286557.1 | NP_001273486.1 | 6 - 7 | 1003 | 55 | ||
| NM_001286558.1 | NP_001273487.1 | 5 - 6 | 934 | 55 | ||
| GenBank mRNA | AB057423.1 | - | 5 - 6 | 907 | 55 | |
| AF329359.1 | - | 5 - 6 | 944 | 55 | ||
| AF329360.1 | - | 6 - 7 | 1013 | 55 | ||
| AK161511.1 | - | 5 - 6 | 920 | 55 | ||
| BC024377.1 | - | 5 - 6 | 895 | 4976492 | 55 | |
| U67176.1 | - | 7 - 8 | 936 | 55 | ||
| U77383.1 | - | 5 - 6 | 556 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtl5
Genomic Map