Assay Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039657.2 | NP_001034746.1 | 7 - 8 | 1155 | 140 | |
| NM_001286557.1 | NP_001273486.1 | 8 - 9 | 1239 | 140 | ||
| NM_001286558.1 | NP_001273487.1 | 7 - 8 | 1170 | 140 | ||
| GenBank mRNA | AB057423.1 | - | 7 - 8 | 1143 | 140 | |
| AF329359.1 | - | 7 - 8 | 1180 | 140 | ||
| AF329360.1 | - | 8 - 9 | 1249 | 140 | ||
| AK015724.1 | - | 7 - 8 | 1095 | 140 | ||
| AK161511.1 | - | 7 - 8 | 1156 | 140 | ||
| BC024377.1 | - | 7 - 8 | 1131 | 4976492 | 140 | |
| U67176.1 | - | 9 - 10 | 1172 | 140 | ||
| U77383.1 | - | 7 - 8 | 792 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtl5
Genomic Map