Assay Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039657.2 | NP_001034746.1 | 8 - 9 | 1333 | 73 | |
| NM_001286557.1 | NP_001273486.1 | 9 - 10 | 1417 | 73 | ||
| NM_001286558.1 | NP_001273487.1 | 8 - 9 | 1348 | 73 | ||
| GenBank mRNA | AB057423.1 | - | 8 - 9 | 1321 | 73 | |
| AF329359.1 | - | 8 - 9 | 1358 | 73 | ||
| AF329360.1 | - | 9 - 10 | 1427 | 73 | ||
| AK015724.1 | - | 8 - 9 | 1273 | 73 | ||
| AK161511.1 | - | 8 - 9 | 1334 | 73 | ||
| BC024377.1 | - | 8 - 9 | 1309 | 4976492 | 73 | |
| U67176.1 | - | 10 - 11 | 1350 | 73 | ||
| U77383.1 | - | 8 - 9 | 970 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | U77383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | AK161511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | BC024377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | U67176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | AK015724.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | U77383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | AK161511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | BC024377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | U67176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177271 | AK015724.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060532 | AK015732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060532 | NM_001039658.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102456 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137598 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127891 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | U77383.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | AK161511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | BC024377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | U67176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | AK015724.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | U77383.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | AK161511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | BC024377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | U67176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177271 | AK015724.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060532 | AK015732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060532 | NM_001039658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P454127 | AK053809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362423 | U77383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362423 | AK161511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362423 | U67176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362423 | AB057423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362423 | NM_001039657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | U77383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | AK161511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | BC024377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | U67176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | AK015724.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | U77383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | AK161511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | BC024377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | U67176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177271 | AK015724.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14142352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14142350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14142349 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14142355 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14503421 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460191 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460181 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425283_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443304_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443831_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431709_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460177 | - | - |
| Mouse Genome 430 2.0 Array | 1431709_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431709_at | - | - |
| Mouse Genome 430 2.0 Array | 1443831_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1443304_at | - | - |
| Mouse Genome 430 2.0 Array | 1443831_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1425283_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425283_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1443304_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17771_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17771_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000077.mm.1 | NM_001039657.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000077.mm.1 | NM_001286557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000077.mm.1 | NM_001286558.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000077.mm.1 | TC1900000011.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000075.mm.1 | NM_001039657.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000075.mm.1 | NM_001286557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000075.mm.1 | NM_001286558.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000075.mm.1 | TC1900000011.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000074.mm.1 | NM_001039657.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000074.mm.1 | NM_001286557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000074.mm.1 | NM_001286558.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000074.mm.1 | TC1900000011.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000076.mm.1 | NM_001039657.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000076.mm.1 | NM_001286557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000076.mm.1 | NM_001286558.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000076.mm.1 | TC1900000011.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425283_a_at | - | - |
| Mouse Expression Set 430 | 1425283_PM_a_at | - | - |
| Mouse Expression Set 430 | 1443304_at | - | - |
| Mouse Expression Set 430 | 1431709_PM_at | - | - |
| Mouse Expression Set 430 | 1443831_PM_s_at | - | - |
| Mouse Expression Set 430 | 1431709_at | - | - |
| Mouse Expression Set 430 | 1443304_PM_at | - | - |
| Mouse Expression Set 430 | 1443831_s_at | - | - |
| Murine Genome U74v2 Set | 167399_PM_r_at | - | - |
| Murine Genome U74v2 Set | 167399_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425283_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425283_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425283_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425283_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431709_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443831_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443304_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443831_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443304_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431709_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104050301 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6940300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104050301 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2602024 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2602024 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217046 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104050301 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104050301 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2602024 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3119327 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3119327 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217046 | - | - |

Mtl5
Genomic Map