Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900000018.mm.1 |
NM_172252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000018.mm.1 |
NM_172252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000018.mm.1 |
TC1900000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000013.mm.1 |
NM_172252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000013.mm.1 |
NM_172252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000013.mm.1 |
TC1900000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000014.mm.1 |
NM_172252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000014.mm.1 |
NM_172252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000014.mm.1 |
TC1900000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000020.mm.1 |
NM_172252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000020.mm.1 |
NM_172252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000020.mm.1 |
TC1900000005.mm.1 |
MTA TC |