Assay Details
| Gene Symbol: Mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172252.3 | NP_758456.2 | 3 - 4 | 272 | 60 | |
| GenBank mRNA | AK003285.1 | - | 3 - 4 | 259 | 60 | |
| AK012784.1 | - | 3 - 4 | 264 | 60 | ||
| AK020739.1 | - | 3 - 4 | 273 | 60 | ||
| BC028768.1 | - | 3 - 4 | 247 | 1282313 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl21
Genomic Map